Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Cytochalasin E

Cytochalasin E

Supplier:   CAYMAN
Cytochalasin E
Supplier:  CAYMAN
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:36011-19-5
  • Formula:C₂₈H₃₃NO₇
  • Molecular Weight:495.57 g/mol
  • Melting Pt:206…208 °C
  • EINECS:252-835-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 120,43 €
    120,43 €
    1 * 1 mg
       120,43 €

  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 474,31 €
    474,31 €
    1 * 5 mg
       474,31 €

  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 884,92 €
    884,92 €
    1 * 10 mg
       884,92 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.