Order Entry
Slovakia
ContactUsLinkComponent
 
 
 
Hydrochloric acid 35% Ph. Eur., extra pure
  SCARAC07561000
 :  SCHARLAB
 :  
 
 
Hydrochloric acid 35% Ph. Eur., extra pure
  SCARAC07561000
 :  SCHARLAB
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :
 :110 °C (1013 hPa)
 :–30 °C
 :1.18 g/cm³ (20 °C)
 :8,II
 :36.46 g/mol
 :231-595-7
 :1789
 :GHS05
 :GHS07
 :Ambient
 :7647-01-0
 :HCl
 :MFCD00011324