Order Entry
Slovakia
ContactUsLinkComponent
1-Octanesulphonic acid sodium salt monohydrate for HPLC
1-Octanesulphonic acid sodium salt monohydrate for HPLC
Catalog # SCARAC17020025
Supplier:  SCHARLAB
CAS Number:  
1-Octanesulphonic acid sodium salt monohydrate for HPLC
Catalog # SCARAC17020025
Supplier:  SCHARLAB
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :234.29 g/mol
Storage Temperature:Ambient
EINECS:226-195-4
CAS Number:207596-29-0
Formula:C₈H₁₇NaO₃S·1H₂O
MDL Number:MFCD00007544