1-Hexanesulphonic acid sodium salt monohydrate for HPLC
Catalog # SCARAC12470025
Supplier: SCHARLAB
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:206.24 g/mol
Storage Temperature:Ambient
EINECS:220-601-3
CAS Number:207300-91-2
Formula:C₆H₁₃NaO₃S·1H₂O
MDL Number:MFCD00149549