Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Dichlorprop
Supelco® PESTANAL® Organic Reference Standard, Dichlorprop
  45436-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dichlorprop
  45436-250MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    120-36-5
  • Description:
    Dichlorprop
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H312 - H315 - H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002645
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-(2,4-Dichlorophenoxy)propionic acid
  • Molecular weight:
    235,06
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS05, GHS07
 : P280 - P305 + P351 + P338