Order Entry
Slovakia
ContactUsLinkComponent
N(α)-Boc-L-tryptophan ≥98%
N(α)-Boc-L-tryptophan ≥98%
Katalógové číslo ABCRAB114355.0005
Supplier:  abcr
CAS Number:  
N(α)-Boc-L-tryptophan ≥98%
Katalógové číslo ABCRAB114355.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :304.3 g/mol
Melting Pt:∼ 138 °C (dec.)
Storage Temperature:Refrigerator
EINECS:236-072-7
CAS Number:13139-14-5
Formula:C₁₆H₂₀N₂O₄
MDL Number:MFCD00065595