Pharmaceutical secondary standard, Metformin hydrochloride, (CRM), Supelco®
PHR1084-500MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:500 mg
- CAS No.:1115-70-4
- Melting point:223-226 °C (lit.)
- Formula:NH₂C(=NH)NHC(=NH)N(CH₃)₂ · HCl
- Description:Metformin hydrochloride
- Form:Neat
Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity.
- Traceable to PhEur M0605000
- Traceable to USP 1396309
Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.
: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).