

Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid
Catalog # SCARAC17651000
Supplier: SCHARLAB
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8,II
:GHS02
:GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325