2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulphonamide
Catalog # J65391.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
Specifications
About this item
CAS Number:313981-55-4
:
Formula:C₁₆H₁₆F₃NO₂S
MDL Number:MFCD00784362
:343.36 g/mol
Storage Temperature:Ambient