Safety ankle boots, lace-up, Steelite™ WORK Constructo Nubuck, FW32
Supplier: Portwest
Certificates
About this item
Anti-static ankle boots made from water resistant nubuck full grain leather with a dual-density phylon/rubber sole resistant to oil and heat (up to 300 °C).
- Shock absorber heel
- Protective steel toecap (200 J)
- Puncture resistant steel midsole
Colour: Black/honey.
Colour: Black/honey
Certifications: EN ISO 20345:2011; S3, SRA.
Frequently Bought Together