Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lead nitrate solution
Lead nitrate solution Reag. Ph. Eur. 1048301
Health
Environment
Lead nitrate solution Reag. Ph. Eur. 1048301
Catalog # 87861.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Health
Environment
Lead nitrate solution Reag. Ph. Eur. 1048301
Catalog # 87861.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
UN:
3082
ADR:
9,III
:
GHS08
:
GHS09
Product Details
Documentation