3,5-Dinitrosalicylic acid
Catalog # LACH30540
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:228.12 g/mol
Melting Pt:170…174 °C
Storage Temperature:Ambient
EINECS:210-204-3
UN:2811
ADR:6.1,III
:GHS07
CAS Number:609-99-4
Formula:C₇H₄N₂O₇
MDL Number:MFCD00007104