Order Entry
Slovakia
ContactUsLinkComponent
Trichloroacetic acid (TCA)
Corrosive
Irritant
Environment
Trichloroacetic acid (TCA)
Catalog # LACH30621
Supplier:  LACH NER
CAS Number:  
undefined
Corrosive
Irritant
Environment
Trichloroacetic acid (TCA)
Catalog # LACH30621
Supplier:  LACH NER
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
EINECS:200-927-2
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:76-03-9
Formula:Cl₃CCOOH
MDL Number:MFCD00004177