Order Entry
Slovakia
ContactUsLinkComponent
Sodium hydroxide 1 mol/l (1 N) in aqueous solution
Corrosive
Sodium hydroxide 1 mol/l (1 N) in aqueous solution
Catalog # LACH61009001P0000
Supplier:  LACH NER
CAS Number:  
Corrosive
Sodium hydroxide 1 mol/l (1 N) in aqueous solution
Catalog # LACH61009001P0000
Supplier:  LACH NER
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:3266
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548