Murexide
Supplier: LACH NER
Certificates
About this item
CAS Number:3051-09-0
:
Formula:C₈H₈N₆O₆
MDL Number:MFCD00012777
:284.19 g/mol
Merck Index:13,06330
Melting Pt:300 °C
Storage Temperature:Ambient
EINECS:221-266-6
Frequently Bought Together