Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution
Catalog # LACH61009100P0000
Supplier: LACH NER
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548