
Potassium thiocyanate
Catalog # LACH30890
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:175 °C
Density:1.61 g/cm³ (20 °C)
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
:GHS07
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413