Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Naphthalene

Naphthalene

Plastic bottle for solids, 500 g
Catalog # LACH40104AP0G0500
Health
Irritant
Environment
Supplier:   LACH NER
Naphthalene
Catalog # LACH40104AP0G0500
Supplier:  LACH NER
CAS Number:  91-20-3
Naphthalene
Naphthalene

Plastic bottle for solids, 500 g

CAS Number:  91-20-3
Price
16,77 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:91-20-3
  • Formula:C₁₀H₈
  • MDL Number:MFCD00001742
  • Molecular Weight:128.17 g/mol
  • Merck Index:12,06457
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:79…82 °C
  • Density:1.145 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Ambient
  • EINECS:202-049-5
  • UN:1334
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications