Disodium hydrogen phosphate dodecahydrate
Supplier: LACH NER
Certificates
About this item
CAS Number:10039-32-4
:
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181
:358.14 g/mol
Merck Index:13,08733
Melting Pt:35 °C
Density:1.52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
Frequently Bought Together