Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium oxide
Magnesium oxide light
Magnesium oxide light
Magnesium oxide light
LACH30135AP0G0150
:
LACH NER
:
Magnesium oxide light
LACH30135AP0G0150
:
LACH NER
:
Pack type:
Plastic bottle for solids
Pk:
100 g
Seq:
3401
:
:
40.3 g/mol
:
3600 °C (1013 hPa)
:
2800 °C
:
3.58 g/cm³ (25 °C)
:
215-171-9
:
1309-48-4
:
MgO
:
MFCD00011109