Isoamyl alcohol
Catalog # LACH30464
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,05214
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
EINECS:204-633-5
UN:1105
ADR:3,III
:GHS02
:GHS07
CAS Number:123-51-3
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934