D-(-)-Mannitol
Supplier: LACH NER
Certificates
About this item
CAS Number:69-65-8
:
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287
:182.17 g/mol
Merck Index:13,05769
Boiling Pt:290…295 °C (4 hPa)
Melting Pt:164…169 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-711-8
Frequently Bought Together