

Copper(II) sulphate
Catalog # LACH30181AP0G10001
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
Specifications
About this item
:
:159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
:GHS07
:GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981