Ammonium dihydrogen phosphate
Catalog # LACH30144
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:13,00547
Melting Pt:190 °C
Density:1.8 g/cm³ (20 °C)
:115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396