
Potassium hexacyanoferrate(II) trihydrate
Supplier: LACH NER
Certificates
About this item
CAS Number:14459-95-1
:
Formula:K₄Fe(CN)₆·3H₂O
MDL Number:MFCD00167023
:422.39 g/mol
Merck Index:12,07793
Melting Pt:70 °C
Density:1.889 g/cm³ (20 °C)
EINECS:237-722-2
:GHS09
Storage Temperature:Ambient
Frequently Bought Together