Urea
LACH40096CP0G1000
: LACH NER
:
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
:
:13,09935
:197 °C (1013 hPa)
:132.7 °C
:1.34 g/cm³ (20 °C)
:60.06 g/mol
:Ambient
:200-315-5
:01-2119463277-33
:57-13-6
:NH₂CONH₂
:MFCD00008022