Order Entry
Slovakia
ContactUsLinkComponent
 
Bromoethylene 1 mol/l in THF, AcroSeal®
Catalog # 428771000.
CAS Number:  
undefined
Bromoethylene 1 mol/l in THF, AcroSeal®
Catalog # 428771000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    0.90 to 1.10 M
  • CO2 absorption capacity:
    ≥19.0 %

Specification Test Results

About this item

 :
 :106.95 g/mol
Flash Pt:–17 °C
Storage Temperature:Refrigerator
EINECS:209-800-6
UN:1993
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
CAS Number:593-60-2
Formula:C₂H₃Br
MDL Number:MFCD00000183