Orthoboric acid
Supplier: LACH NER
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
Frequently Bought Together