Order Entry
Slovakia
ContactUsLinkComponent
Sodium hydroxide, white pearls ACS, ISO for analysis
Sodium hydroxide, white pearls ACS, ISO for analysis
Supplier:  LACH NER
Certificates
Sodium hydroxide, white pearls ACS, ISO for analysis
Supplier:  LACH NER

Specification Test Results

  • Appearance:
    White pearls
  • Assay:
    98.0 - 101.0 %
  • Carbonates (Na₂CO₃):
    max. 1.0 %
  • Total nitrogen (N):
    max. 0.0005 %

Specification Test Results

Frequently Bought Together