Order Entry
Slovakia
ContactUsLinkComponent
Periodic acid
Oxidizing
Corrosive
Periodic acid
Catalog # LACH10036AP0G0100
Supplier:  LACH NER
CAS Number:  
undefined
Oxidizing
Corrosive
Periodic acid
Catalog # LACH10036AP0G0100
Supplier:  LACH NER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :227.94 g/mol
Merck Index:13,07248
Melting Pt:122 °C
Density:1.3875 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-937-0
UN:3085
ADR:5.1,I
 :GHS03
 :GHS05
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344