Sodium chloride
Supplier: LACH NER
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Frequently Bought Together