Sodium bromide
Catalog # LACH30006CP0G0500
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:102.89 g/mol
Merck Index:13,08666
Boiling Pt:1396 °C (1013 hPa)
Melting Pt:753 °C
Density:3.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-599-9
CAS Number:7647-15-6
Formula:NaBr
MDL Number:MFCD00003475