Order Entry
Slovakia
ContactUsLinkComponent
n-Hexane
Flammable
Health
Irritant
Environment
n-Hexane
Catalog # LACH20031AT0M1000
Supplier:  LACH NER
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Hexane
Catalog # LACH20031AT0M1000
Supplier:  LACH NER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520