Chloroform
Glass bottle, 2,5 L
LACH20034LT1M2500
: LACH NER
: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- :67-66-3
- :CHCl₃
- :MFCD00000826
- :119.38 g/mol
- :13,02160
- :61.7 °C (1013 hPa)
- :–63 °C
- :1.4832 g/cm³ (20 °C)
- :Ambient
- :200-663-8
- :1888
- :6.1,III
- :05-2114614906-45
- Pack type:Glass bottle
- Pk:2,5 L