Order Entry
Slovakia
ContactUsLinkComponent
Potassium acetate
  LACH40113CP0G10001
 :  LACH NER
 :  
undefined
Potassium acetate
  LACH40113CP0G10001
 :  LACH NER
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    1.000 g
  • Seq:
    3047
 :
 :98.14 g/mol
 :13,07687
 :304 °C
 :1.57 g/cm³ (25 °C)
 :>250 °C
 :Ambient
 :204-822-2
 :127-08-2
 :CH₃COOK
 :MFCD00012458