Acetone
Supplier: LACH NER
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95.4 °C
- Density:0.792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
3 Options Available Below
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95.4 °C
- Density:0.792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- LACH20001CT0M1000
- LACH20001CT0M2500
- LACH20001CT0G7900
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.LACH20001CT0M1000Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health - Specifications:
More
Specifications
Cat. No.LACH20001CT0M2500Environmentally PreferableEnvironmental certification - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.LACH20001CT0G7900Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



