
Calcium chloride dihydrate
Supplier: LACH NER
Certificates
About this item
CAS Number:10035-04-8
:
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613
:147.01 g/mol
Merck Index:13,01660
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1.85 g/cm³ (20 °C)
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together