Barium chloride dihydrate
LACH30072APOG0100
: LACH NER
:
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
:
:244.27 g/mol
:14,00971
:1560 °C (1013 hPa)
:960 °C
:3.86 g/cm³ (25 °C)
:Ambient
:233-788-1
:1564
:6.1,III
:GHS06
:10326-27-9
:BaCl₂·2H₂O
:MFCD00149154