Sodium chromate tetrahydrate
Catalog # LACH30110AP0G0100
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:234.03 g/mol
Melting Pt:792.2 °C
Density:2.73 g/cm³ (20 °C)
EINECS:231-889-5
UN:3288
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:10034-82-9
Formula:CrNa₂O₄·4H₂O
MDL Number:MFCD00149167