



Mercury(II) chloride
Supplier: LACH NER
Certificates
About this item
CAS Number:7487-94-7
:
Formula:HgCl₂
MDL Number:MFCD00011041
:271.5 g/mol
Merck Index:13,05901
Boiling Pt:302 °C (1013 hPa)
Melting Pt:273 °C
Density:5.44 g/cm³ (20 °C)
EINECS:231-299-8
UN:1624
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together