

Isoamyl alcohol
LACH20035AT0M1000
: LACH NER
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
:
:88.15 g/mol
:13,05214
:130 °C (1013 hPa)
:–117 °C
:0.812 g/cm³ (20 °C)
:42 °C
:Ambient
:204-633-5
:1105
:3,III
:GHS02
:GHS07
:123-51-3
:(CH₃)₂CHCH₂CH₂OH
:MFCD00002934