Order Entry
Slovakia
ContactUsLinkComponent
Ammonium sulphate
Ammonium sulphate
Catalog # LACH30165CP0G1000
Supplier:  LACH NER
CAS Number:  
Ammonium sulphate
Catalog # LACH30165CP0G1000
Supplier:  LACH NER
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1.000 g
  • Seq:
    3047

Specifications

About this item

 :
 :132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391