Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Ammonium peroxodisulphate (APS)

Ammonium peroxodisulphate (APS)

Plastic bottle for solids, 500 g
Catalog # LACH30154CP0G0500
Oxidizing
Health
Irritant
Supplier:   LACH NER
Ammonium peroxodisulphate (APS)
Catalog # LACH30154CP0G0500
Supplier:  LACH NER
CAS Number:  7727-54-0
Ammonium peroxodisulphate (APS)
Ammonium peroxodisulphate (APS)

Plastic bottle for solids, 500 g

CAS Number:  7727-54-0
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Peroxidisulphuric acid diammonium salt, Ammonium persulphate
  • CAS Number:7727-54-0
  • Formula:(NH₄)₂S₂O₈
  • MDL Number:MFCD00003390
  • Molecular Weight:228.2 g/mol
  • Merck Index:13,00545
  • Melting Pt:120 °C
  • Density:1.98 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-786-5
  • UN:1444
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications