Diethylene glycol
Catalog # LACH20017CT0M10001
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:106.12 g/mol
Merck Index:13,03146
Boiling Pt:245 °C (1013 hPa)
Melting Pt:–10 °C
Density:1.118 g/cm³ (20 °C)
Flash Pt:123 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-872-2
:GHS07
CAS Number:111-46-6
Formula:C₄H₁₀O₃
MDL Number:MFCD00002882