Dimethyl sulphoxide
LACH20022CT0M1000
: LACH NER
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
:
:78.14 g/mol
:13,03285
:189 °C (1013 hPa)
:18.5 °C
:1.101 g/cm³ (20 °C)
:87 °C
:Ambient
:200-664-3
:67-68-5
:(CH₃)₂SO
:MFCD00002089