

Copper(II) chloride dihydrate
Catalog # LACH30089AP0G0100
Supplier: LACH NER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C
Density:2.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS07
:GHS09
CAS Number:10125-13-0
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674