

Copper(II) chloride dihydrate
Supplier: LACH NER
Certificates
About this item
CAS Number:10125-13-0
:
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674
:170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C
Density:2.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS07
:GHS09
Frequently Bought Together