Supelco® Organic Reference Standard, Ginkgolide C
18309-10MG-F
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:15291-76-6
- Description:Ginkgolide C
- Seq:2275
- Storage conditions:–20 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD02094178
- M-Clarity quality level:MQ100
- Synonym(s):1,7-Dihydroxy-ginkgolide A
- Molecular weight:440,40
- Flash point:Not applicable
: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338