Supelco® Organic Reference Standard, Ginkgolide B from Ginkgo biloba leaves
Supplier: Merck
Certificates
About this item
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
Specifications
- CAS No.:15291-77-7
- Storage conditions:–20 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD05662347
- M-Clarity quality level:MQ100
- Synonym(s):BN-52021
- Molecular weight:424,40
Specifications
Frequently Bought Together