Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Ginkgolide A from Ginkgo biloba leaves
Supelco® Organic Reference Standard, Ginkgolide A from Ginkgo biloba leaves
Catalog # G4028-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ginkgolide A from Ginkgo biloba leaves
Catalog # G4028-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard, ≥90% (HPLC)
  • CAS No.:
    15291-75-5
  • Description:
    Ginkgolide A from Ginkgo biloba leaves
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00133365
  • Molecular weight:
    408,40

Specifications

About this item

Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338